This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495899_m1
See other UCHL1 GE Assays ›
Gene Symbol:
Uchl1
Gene Name:
ubiquitin carboxy-terminal hydrolase L1
Gene Aliases:
AW822034, C88048, PGP 9.5, PGP9.5, R75593, UCH-L1, UCHL-1, gad
Chromosome Location:
Chr.5: 66676121 - 66687234 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495899_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uchl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011670.2 NP_035800.2 3 - 4 349 73
GenBank mRNA AB025313.1 - 3 - 4 200 73
AF172334.1 - 3 - 4 188 73
AK004066.1 - 3 - 4 226 73
AK013729.1 - 3 - 4 229 73
AK170728.1 - 1 - 2 187 73
BC039177.1 - 3 - 4 215 4506229 73
CX241178.1 - 3 - 4 349 73
JX050004.1 - 1 - 2 153 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000640.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uchl1

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

response to ischemia
proteolysis
ubiquitin-dependent protein catabolic process
axonogenesis
axon target recognition
adult walking behavior
cell proliferation
protein deubiquitination
sensory perception of pain
axonal transport of mitochondrion
eating behavior
negative regulation of MAP kinase activity
muscle fiber development
neuromuscular process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
omega peptidase activity
hydrolase activity
ligase activity
ubiquitin protein ligase binding
alpha-2A adrenergic receptor binding
ubiquitin binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500005627.mm.1 NM_011670.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005627.mm.1 NM_011670.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500005627.mm.1 TC0500000640.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500005622.mm.1 NM_011670.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005622.mm.1 NM_011670.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500005622.mm.1 TC0500000640.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500005620.mm.1 NM_011670.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005620.mm.1 NM_011670.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500005620.mm.1 TC0500000640.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500005626.mm.1 NM_011670.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005626.mm.1 NM_011670.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500005626.mm.1 TC0500000640.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 926789 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0