This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494740_g1
See other GRB10 GE Assays ›
Gene Symbol:
Grb10
Gene Name:
growth factor receptor bound protein 10
Gene Aliases:
5730571D09Rik, AI325020, Meg1, mKIAA0207
Chromosome Location:
Chr.11: 11930499 - 12037422 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494740_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grb10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177629.1 NP_001171100.1 8 - 9 1003 97
NM_010345.4 NP_034475.2 9 - 10 1328 97
GenBank mRNA AF022072.1 - 9 - 10 1302 97
AK030727.1 - 9 - 10 1263 97
AK032927.1 - 9 - 10 1309 97
AK163841.1 - 9 - 10 1309 97
AK169000.1 - 3 - 4 292 97
BC016111.1 - 8 - 9 1003 4481345 97
BC053842.1 - 9 - 10 1295 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002227.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grb10

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
insulin receptor signaling pathway
positive regulation of signal transduction
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
response to insulin
negative regulation of phosphorylation
positive regulation of phosphorylation
negative regulation of glycogen biosynthetic process
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
phosphotyrosine binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100021618.mm.1 NM_001177629.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021618.mm.1 NM_001177629.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021618.mm.1 NM_010345.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021618.mm.1 NM_010345.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100021618.mm.1 TC1100002227.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100021619.mm.1 NM_001177629.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021619.mm.1 NM_001177629.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021619.mm.1 NM_010345.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021619.mm.1 NM_010345.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100021619.mm.1 TC1100002227.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0