Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_001190703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_001190703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_001190704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_001190704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_001190705.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_001190705.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_010052.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
NM_010052.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007991.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007987.mm.1 |
NR_033813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007987.mm.1 |
NR_033813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007987.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007988.mm.1 |
NR_033813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007988.mm.1 |
NR_033813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007988.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_001190703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_001190703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_001190704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_001190704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_001190705.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_001190705.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_010052.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NM_010052.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NR_033813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
NR_033813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007993.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_001190703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_001190703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_001190704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_001190704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_001190705.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_001190705.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_010052.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
NM_010052.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007989.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_001190703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_001190703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_001190704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_001190704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_001190705.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_001190705.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_010052.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
NM_010052.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007992.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_001190703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_001190703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_001190704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_001190704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_001190705.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_001190705.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_010052.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NM_010052.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NR_033813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
NR_033813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007995.mm.1 |
TC1200001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_001190703.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_001190703.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_001190704.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_001190704.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_001190705.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_001190705.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_010052.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NM_010052.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NR_033813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
NR_033813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007998.mm.1 |
TC1200001125.mm.1 |
MTA TC |