This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00493644_m1
See other TGFBI GE Assays ›
Gene Symbol:
Tgfbi
Gene Name:
transforming growth factor, beta induced
Gene Aliases:
68kDa, AI181842, AI747162, Beta-ig, big-h3
Chromosome Location:
Chr.13: 56609603 - 56639339 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00493644_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tgfbi
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009369.4 NP_033395.1 11 - 12 1591 121
GenBank mRNA AK084431.1 - 11 - 12 1609 121
AK135536.1 - 11 - 12 1636 121
AK142323.1 - 11 - 12 1611 121
AK150449.1 - 11 - 12 1609 121
AK150833.1 - 11 - 12 1609 121
AK150969.1 - 11 - 12 1610 121
AK151680.1 - 11 - 12 1606 121
AK152365.1 - 11 - 12 1610 121
AK152662.1 - 11 - 12 1609 121
AK155572.1 - 11 - 12 1606 121
AK155828.1 - 11 - 12 1606 121
AK159711.1 - 11 - 12 1605 121
AK164373.1 - 11 - 12 1609 121
AK169912.1 - 11 - 12 1611 121
AK170495.1 - 11 - 12 1606 121
AK171557.1 - 10 - 11 1430 121
BC129900.1 - 11 - 12 1584 40111549 121
BC129901.1 - 11 - 12 1584 40111550 121
L19932.1 - 11 - 12 1592 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000691.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tgfbi

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
extracellular matrix organization
collagen binding
cell adhesion molecule binding
extracellular matrix binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0