This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00492979_g1
See other KRT16 GE Assays ›
Gene Symbol:
Krt16
Gene Name:
keratin 16
Gene Aliases:
AI324768, K16, Krt1-16
Chromosome Location:
Chr.11: 100246091 - 100248962 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00492979_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Krt16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008470.1 NP_032496.1 7 - 8 1330 87
GenBank mRNA AF053235.1 - 7 - 8 1330 87
AK137631.1 - 7 - 8 1394 87
BC103615.1 - 7 - 8 1333 40046209 87
BC103616.1 - 7 - 8 1333 40046211 87
BC103617.1 - 7 - 8 1333 40046210 87
BC103666.1 - 7 - 8 1330 40046212 87
BC114977.1 - 7 - 8 1330 40046220 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003762.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Krt16

Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
immune system process
inflammatory response
aging
keratinocyte differentiation
negative regulation of cell migration
keratinization
hair cycle
innate immune response
intermediate filament cytoskeleton organization
keratinocyte migration
establishment of skin barrier
structural molecule activity
structural constituent of cytoskeleton
protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007756 AK137631.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007756 NM_008470.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007756 AF053235.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007756 AK137631.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007756 NM_008470.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007756 AF053235.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P457747 NM_008470.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P457747 AF053235.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067005 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067006 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067012 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066990 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066987 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066983 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067007 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067002 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066989 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067014 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066976 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066977 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066992 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066978 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067009 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067001 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28067003 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066995 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066998 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066971 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066994 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066952 - -
Whole Mouse Genome G4122A A_51_P457747 NM_008470.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P457747 AF053235.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111744 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09432650 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09432654 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09432651 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09432653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067027 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066989 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067053 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066939 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067036 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066977 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066951 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066943 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066998 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067047 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066994 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28067003 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067014 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066946 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067012 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066994 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066968 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067019 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066971 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066954 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09432630 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09484142 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066990 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066978 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067024 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066950 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067009 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09484153 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067030 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066998 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067027 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066937 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09484140 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09432650 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067007 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066936 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067032 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067017 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067022 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066996 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066992 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28067002 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09685850 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09432643 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066988 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066982 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09484125 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066999 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066939 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066951 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09484145 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28066948 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10391065 - -
Affymetrix Mouse Gene 1.0 ST Array 10391063 - -
Affymetrix Mouse Gene 1.0 ST Array 10391062 - -
Affymetrix Mouse Gene 1.0 ST Array 10391064 - -
Affymetrix HT MG-430 PM Array Plate 1448932_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10391061 - -
Mouse Genome 430 2.0 Array 1448932_PM_at - -
Mouse Genome 430 2.0 Array 1448932_at - -
Affymetrix Mouse Clariom S Array PSR1100034525.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034525.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034525.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034520.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034520.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034520.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034529.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034529.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034529.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034526.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034526.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034526.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034521.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034521.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034521.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034527.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034527.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034527.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034524.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034524.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034524.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034522.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034522.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034522.mm.1 TC1100003762.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100034528.mm.1 NM_008470.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034528.mm.1 NM_008470.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034528.mm.1 TC1100003762.mm.1 MTA TC
Mouse Expression Set 430 1448932_at - -
Mouse Expression Set 430 1448932_PM_at - -
Murine Genome U74v2 Set 103589_at - -
Murine Genome U74v2 Set 103589_PM_at - -
Mouse Genome 430A 2.0 Array 1448932_at - -
Mouse Genome 430A 2.0 Array 1448932_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1448932_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1448932_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 1400551 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001400551 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229813 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1229813 - -
mouse-6 v1.1 (Illumina) scl39569.8.1_38-S - -
Illumina Sentrix mouse-6.v1 bead array 1400551 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl39569.8.1_38-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400551 - -
MouseRef-8 v2 Expression BeadChip ILMN_1229813 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229813 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0