This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00491649_m1
See other SCUBE1 GE Assays ›
Gene Symbol:
Scube1
Gene Name:
signal peptide, CUB domain, EGF-like 1
Gene Aliases:
7330410C13Rik, A630023E24Rik
Chromosome Location:
Chr.15: 83602583 - 83725049 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00491649_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Scube1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271472.1 NP_001258401.1 16 - 17 2115 74
NM_001271473.1 NP_001258402.1 13 - 14 1782 74
NM_022723.3 NP_073560.2 15 - 16 2025 74
GenBank mRNA AF276425.1 - 15 - 16 1892 74
AK041590.1 - 15 - 16 2008 74
BC066066.1 - 16 - 17 2073 6850999 74
BC080278.1 - 13 - 14 1750 30354974 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Scube1

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

protein homooligomerization
calcium ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0