This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00491481_g1
See other CENPH GE Assays ›
Gene Symbol:
Cenph
Gene Name:
centromere protein H
Gene Aliases:
1700021I11Rik, 2410018A12Rik, 2610042E16Rik, 2810046K12Rik, AU044255, CENP-H, ENP
Chromosome Location:
Chr.13: 100759686 - 100775899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00491481_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cenph
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021886.1 NP_068686.1 3 - 4 316 140
GenBank mRNA AB017634.1 - 3 - 4 316 140
AK006208.1 - 3 - 4 300 140
AK011264.1 - 3 - 4 301 140
AK011738.1 - 3 - 4 301 140
BC025084.1 - 3 - 4 267 3501244 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002542.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cenph

Gene Ontology Categories:

Function(s) Process(es)

chromosome segregation
mitotic nuclear division
kinetochore assembly
kinetochore organization
kinetochore binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017326.mm.1 NM_021886.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017326.mm.1 TC1300002542.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0