This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00491393_m1
See other SLC4A8 GE Assays ›
Gene Symbol:
Slc4a8
Gene Name:
solute carrier family 4 (anion exchanger), member 8
Gene Aliases:
AW493845, C230026C11, NDCBE, kNBC-3
Chromosome Location:
Chr.15: 100761700 - 100823971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00491393_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021530.2 NP_067505.2 1 - 2 225 68
GenBank mRNA AF224508.1 - 1 - 2 171 68
AK034237.1 - 1 - 2 226 68
AK039404.1 - 1 - 2 190 68
AK173014.1 - 1 - 2 59 68
BB626223.1 - 1 - 2 226 68
BC030388.1 - 1 - 2 187 5401143 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001054.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a8

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
anion transport
embryo development
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
anion transmembrane transport
transporter activity
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
antiporter activity
anion:anion antiporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008326.mm.1 NM_021530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008326.mm.1 NM_021530.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008326.mm.1 TC1500001054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008329.mm.1 NM_021530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008329.mm.1 NM_021530.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008329.mm.1 TC1500001054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008327.mm.1 NM_021530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008327.mm.1 NM_021530.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008327.mm.1 TC1500001054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008330.mm.1 NM_021530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008330.mm.1 NM_021530.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008330.mm.1 TC1500001054.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 323001 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0