This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00490991_g1
See other SCUBE2 GE Assays ›
Gene Symbol:
Scube2
Gene Name:
signal peptide, CUB domain, EGF-like 2
Gene Aliases:
4932442O19Rik, Cegf1, Cegp1
Chromosome Location:
Chr.7: 109798690 - 109865679 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00490991_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Scube2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020052.2 NP_064436.2 15 - 16 1879 72
GenBank mRNA AK016571.1 - 9 - 10 1026 72
AK142950.1 - 15 - 16 1846 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004084.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Scube2

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700033742.mm.1 NM_020052.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033742.mm.1 NM_020052.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033742.mm.1 TC0700004084.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700033741.mm.1 NM_020052.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033741.mm.1 NM_020052.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033741.mm.1 TC0700004084.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700033743.mm.1 NM_020052.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700033743.mm.1 NM_020052.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700033743.mm.1 TC0700004084.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0