This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00490956_m1
See other MBNL1 GE Assays ›
Gene Symbol:
Mbnl1
Gene Name:
muscleblind-like 1 (Drosophila)
Gene Aliases:
Mbnl, mKIAA0428
Chromosome Location:
Chr.3: 60472791 - 60629750 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00490956_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbnl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253708.1 NP_001240637.1 5 - 6 1851 73
NM_001253710.1 NP_001240639.1 4 - 5 802 73
NM_001253713.1 NP_001240642.1 5 - 6 805 73
NM_020007.3 NP_064391.2 5 - 6 1851 73
GenBank mRNA AB093240.1 - 5 - 6 1855 73
AF231110.1 - 4 - 5 1291 73
AK038242.1 - 4 - 5 886 73
AK153827.1 - 4 - 5 695 73
BC096600.1 - 4 - 5 763 4976244 73
CN663545.1 - 2 - 3 378 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbnl1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
mRNA processing
skeletal muscle tissue development
RNA splicing
regulation of RNA splicing
myoblast differentiation
RNA binding
double-stranded RNA binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0