Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
NM_001253708.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
NM_001253709.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
NM_001253710.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
NM_001253711.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
NM_001253713.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
NM_020007.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003371.mm.1 |
TC0300000485.mm.1 |
MTA TC |