This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00490809_m1
See other GPR132 GE Assays ›
Gene Symbol:
Gpr132
Gene Name:
G protein-coupled receptor 132
Gene Aliases:
G2a
Chromosome Location:
Chr.12: 112850873 - 112868418 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00490809_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019925.4 NP_064309.1 2 - 3 137 73
GenBank mRNA AF083442.1 - 2 - 3 173 73
AK089866.1 - 2 - 3 114 73
AK153765.1 - 2 - 3 138 73
AK154509.1 - 2 - 3 159 73
AK157600.1 - 2 - 3 165 73
BC120522.1 - 1 - 2 64 40129445 73
BC120524.1 - 1 - 2 64 40129447 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002440.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF083442.1,AK089866.1,AK153765.1,BC120522.1,BC120524.1,NM_019925.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr132

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
signal transduction
G-protein coupled receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0