This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00490007_m1
See other CTNNAL1 GE Assays ›
Gene Symbol:
Ctnnal1
Gene Name:
catenin (cadherin associated protein), alpha-like 1
Gene Aliases:
ACRP, AI616177, AW545119, C86009, Catnal1
Chromosome Location:
Chr.4: 56810935 - 56865211 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00490007_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctnnal1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018761.3 NP_061231.3 8 - 9 1286 91
GenBank mRNA AF006071.1 - 8 - 9 1273 91
AK034393.1 - 9 - 10 1286 91
AK038548.1 - 5 - 6 1223 91
AK082755.1 - 8 - 9 1230 91
BC050070.1 - 8 - 9 1263 30012833 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002708.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctnnal1

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
Rho protein signal transduction
cadherin binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400021854.mm.1 NM_018761.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021854.mm.1 NM_018761.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021854.mm.1 TC0400002708.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021856.mm.1 NM_018761.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021856.mm.1 NM_018761.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400021856.mm.1 TC0400002708.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0