This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489499_m1
See other MPP2 GE Assays ›
Gene Symbol:
Mpp2
Gene Name:
membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2)
Gene Aliases:
D11Bwg0652e, Dlg2, Dlgh2, Pals4
Chromosome Location:
Chr.11: 102057016 - 102088636 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489499_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016695.3 NP_057904.1 1 - 2 92 69
GenBank mRNA AF162685.1 - 1 - 2 79 69
AK161577.1 - 1 - 2 85 69
AK172253.1 - 1 - 2 77 69
BC053026.1 - 1 - 2 92 5709543 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003821.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF162685.1,AK161577.1,AK172253.1,BC053026.1,NM_016695.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mpp2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Process(es)

protein binding
PDZ domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100035440.mm.1 NM_016695.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100035440.mm.1 NM_016695.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100035440.mm.1 TC1100003821.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100035442.mm.1 NM_016695.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100035442.mm.1 NM_016695.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100035442.mm.1 TC1100003821.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100035436.mm.1 NM_016695.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100035436.mm.1 NM_016695.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100035436.mm.1 TC1100003821.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0