This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489172_g1
See other CABP2 GE Assays ›
Gene Symbol:
Cabp2
Gene Name:
calcium binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.19: 4081469 - 4087340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489172_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013878.2 NP_038906.2 1 - 2 99 140
GenBank mRNA AF169157.1 - 1 - 2 127 140
BC107029.2 - 1 - 2 99 40050356 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000025.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF169157.1,BC107029.2,NM_013878.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cabp2

Gene Ontology Categories:

Process(es)

calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000307.mm.1 NM_013878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000307.mm.1 NM_013878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000307.mm.1 TC1900000025.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0