This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489010_g1
See other SH3GL1 GE Assays ›
Gene Symbol:
Sh3gl1
Gene Name:
SH3-domain GRB2-like 1
Gene Aliases:
C77078, EEN, SH3P8, Sh3d2b
Chromosome Location:
Chr.17: 56016750 - 56036638 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489010_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sh3gl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013664.3 NP_038692.1 8 - 9 1038 110
GenBank mRNA AK154836.1 - 8 - 9 1037 110
AK162715.1 - 8 - 9 966 110
AK167505.1 - 8 - 9 980 110
BC138602.1 - 8 - 9 977 8861622 110
BC138603.1 - 8 - 9 977 8861623 110
U58885.1 - 8 - 9 977 110

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sh3gl1

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
protein binding
lipid binding
phosphatase binding
identical protein binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0