This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488992_g1
See other MSH5 GE Assays ›
Gene Symbol:
Msh5
Gene Name:
mutS homolog 5 (E. coli)
Gene Aliases:
Mut5
Chromosome Location:
Chr.17: 35028605 - 35048501 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488992_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msh5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF107352.1 - 23 - 24 2356 76
AF146227.1 - 22 - 23 2564 76
AK132626.1 - 10 - 11 1425 76
BC141113.1 - 22 - 23 2484 9055725 76
BC145406.1 - 22 - 23 2474 76
BC145407.1 - 22 - 23 2386 76
CK793627.1 - 1 - 2 215 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001940.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700018384.mm.1 NM_001146215.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700018384.mm.1 NM_013600.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700018384.mm.1 TC1700001940.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0