This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488270_m1
See other SLC12A7 GE Assays ›
Gene Symbol:
Slc12a7
Gene Name:
solute carrier family 12, member 7
Gene Aliases:
AA408796, D13Ertd261e, Kcc4
Chromosome Location:
Chr.13: 73733053 - 73816755 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488270_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc12a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011390.2 NP_035520.1 8 - 9 1193 119
GenBank mRNA AF087436.1 - 8 - 9 1195 119
AK052910.1 - 8 - 9 1244 119
AK083598.1 - 5 - 6 3464 119
AK131129.1 - 8 - 9 1147 119
AK143535.1 - 8 - 9 1187 119
AK149750.1 - 8 - 9 1188 119
AK166215.1 - 8 - 9 1187 119
AK169950.1 - 8 - 9 1183 119
AK171498.1 - 8 - 9 1230 119
BC059242.1 - 8 - 9 1185 119
BC060133.1 - 8 - 9 1185 119
BC079677.1 - 8 - 9 1151 119
BC141107.1 - 8 - 9 1153 9055719 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000887.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc12a7

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
cell volume homeostasis
transmembrane transport
chloride transmembrane transport
symporter activity
cation:chloride symporter activity
potassium:chloride symporter activity
protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300005727.mm.1 NM_011390.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300005727.mm.1 NM_011390.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300005727.mm.1 TC1300000887.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0