This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488239_m1
See other SIN3A GE Assays ›
Gene Symbol:
Sin3a
Gene Name:
transcriptional regulator, SIN3A (yeast)
Gene Aliases:
AW553200, Sin3, mKIAA4126, mSin3A
Chromosome Location:
Chr.9: 57071991 - 57128368 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488239_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sin3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110350.1 NP_001103820.1 3 - 4 652 68
NM_001110351.1 NP_001103821.1 4 - 5 529 68
NM_011378.2 NP_035508.2 4 - 5 760 68
GenBank mRNA AK032209.1 - 3 - 4 520 68
AK211749.1 - 1 - 2 126 68
AK220292.1 - 3 - 4 629 68
BC052716.1 - 3 - 4 539 6837144 68
BC053385.1 - 3 - 4 522 68
BC078454.1 - 3 - 4 513 68
L36831.1 - 3 - 4 443 68
L38620.2 - 2 - 3 419 68
L38621.2 - 2 - 3 419 68
U22394.1 - 4 - 5 768 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000695.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sin3a

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
activation of innate immune response
positive regulation of defense response to virus by host
hematopoietic progenitor cell differentiation
DNA replication
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein deacetylation
aging
response to organonitrogen compound
regulation of hormone levels
positive regulation of G2/M transition of mitotic cell cycle
histone deacetylation
positive regulation of chromatin silencing
cellular protein localization
negative regulation of circadian rhythm
negative regulation of apoptotic process
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
response to methylglyoxal
cellular response to glucose stimulus
negative regulation of protein localization to nucleus
negative regulation of histone H3-K27 acetylation
negative regulation of transcription regulatory region DNA binding
transcription regulatory region sequence-specific DNA binding
RNA polymerase II activating transcription factor binding
RNA polymerase II repressing transcription factor binding
RNA polymerase II transcription corepressor activity
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
RNA binding
histone deacetylase activity
protein binding
transcription factor binding
protein complex binding
protein deacetylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 NM_001110350.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 NM_001110350.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 NM_001110351.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 NM_001110351.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 NM_011378.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 NM_011378.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900005031.mm.1 TC0900000695.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 NM_001110350.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 NM_001110350.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 NM_001110351.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 NM_001110351.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 NM_011378.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 NM_011378.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900005030.mm.1 TC0900000695.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0