This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00487537_g1
See other MAG GE Assays ›
Gene Symbol:
Mag
Gene Name:
myelin-associated glycoprotein
Gene Aliases:
Gma, siglec-4a
Chromosome Location:
Chr.7: 30899175 - 30914922 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00487537_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mag
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010758.2 NP_034888.1 3 - 4 195 86
GenBank mRNA BC101944.1 - 3 - 4 168 40039200 86
BC103521.1 - 3 - 4 168 40039204 86
BC103522.1 - 3 - 4 168 86
BC103523.1 - 3 - 4 168 86
M31811.1 - 3 - 4 195 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002780.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mag

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
negative regulation of neuron projection development
negative regulation of neuron differentiation
positive regulation of astrocyte differentiation
receptor binding
protein binding
protein kinase binding
carbohydrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header