This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486379_m1
See other STRBP GE Assays ›
Gene Symbol:
Strbp
Gene Name:
spermatid perinuclear RNA binding protein
Gene Aliases:
6430510M02Rik, AI852513, C230082I21Rik, C86322, Spnr
Chromosome Location:
Chr.2: 37569868 - 37703969 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486379_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Strbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009261.3 NP_033287.2 9 - 10 1215 91
GenBank mRNA BC014710.1 - 10 - 11 1299 4502840 91
X84692.1 - 1152 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003401.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Strbp

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
multicellular organism development
spermatogenesis
spermatid development
mechanosensory behavior
cell differentiation
DNA binding
RNA binding
double-stranded RNA binding
single-stranded RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200029230.mm.1 NM_009261.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200029230.mm.1 TC0200003401.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0