This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486252_g1
See other SMPD2 GE Assays ›
Gene Symbol:
Smpd2
Gene Name:
sphingomyelin phosphodiesterase 2, neutral
Gene Aliases:
AW108287, nSMase, nSMase1
Chromosome Location:
Chr.10: 41487172 - 41490340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486252_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smpd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009213.2 NP_033239.1 6 - 7 803 77
GenBank mRNA AJ222800.1 - 6 - 7 804 77
AK075616.1 - 6 - 7 813 77
AK088962.1 - 3 - 4 1571 77
BC010978.1 - 6 - 7 813 4193282 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002054.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smpd2

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
response to mechanical stimulus
intracellular signal transduction
positive regulation of apoptotic process
ceramide biosynthetic process
sphingomyelin phosphodiesterase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000014598.mm.1 NM_009213.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014598.mm.1 NM_009213.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000014598.mm.1 TC1000002054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000014597.mm.1 NM_009213.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014597.mm.1 NM_009213.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000014597.mm.1 TC1000002054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0