Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200015346.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015346.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015346.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015355.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015355.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015355.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015345.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015345.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015345.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015349.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015349.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015349.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015356.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015356.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015356.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015353.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015353.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015353.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015344.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015344.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015344.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015352.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015352.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015352.mm.1 |
TC0200001947.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200015354.mm.1 |
NM_009086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015354.mm.1 |
NM_009086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200015354.mm.1 |
TC0200001947.mm.1 |
MTA TC |