Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000000876.mm.1 |
NM_001111065.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000876.mm.1 |
NM_001111065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000876.mm.1 |
NM_009048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000876.mm.1 |
NM_009048.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000876.mm.1 |
TC1000000126.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000000875.mm.1 |
NM_001111065.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000875.mm.1 |
NM_001111065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000875.mm.1 |
NM_009048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000875.mm.1 |
NM_009048.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000000875.mm.1 |
TC1000000126.mm.1 |
MTA TC |