Assay Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009029.2 | NP_033055.2 | 24 - 25 | 2646 | 123 | |
GenBank mRNA | AK132875.1 | - | 13 - 14 | 1392 | 123 | |
AK141028.1 | - | 24 - 25 | 2678 | 123 | ||
AK146467.1 | - | 19 - 20 | 1977 | 123 | ||
BC096525.1 | - | 24 - 25 | 2646 | 6832808 | 123 | |
DQ400415.1 | - | 24 - 25 | 2496 | 123 | ||
M26391.1 | - | 24 - 25 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
