This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00485063_m1
See other MMP15 GE Assays ›
Gene Symbol:
Mmp15
Gene Name:
matrix metallopeptidase 15
Gene Aliases:
AI503551, MT2-MMP
Chromosome Location:
Chr.8: 95352337 - 95374293 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00485063_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmp15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008609.3 NP_032635.1 2 - 3 832 68
GenBank mRNA BC047278.1 - 2 - 3 832 6390113 68
BC057952.1 - 2 - 3 402 5342023 68
D86332.1 - 2 - 3 328 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001133.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mmp15

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
metalloendopeptidase activity
calcium ion binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800008935.mm.1 NM_008609.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800008935.mm.1 TC0800001133.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0