This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00484701_g1
See other GBA GE Assays ›
Gene Symbol:
Gba
Gene Name:
glucosidase, beta, acid
Gene Aliases:
GBA1, GC, GCase, GLUC, betaGC
Chromosome Location:
Chr.3: 89202914 - 89208966 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00484701_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gba
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077411.2 NP_001070879.1 5 - 6 702 65
NM_008094.5 NP_032120.1 6 - 7 871 65
NR_122037.1 6 - 7 871 65
GenBank mRNA AK082767.1 - 5 - 6 700 65
AK152745.1 - 5 - 6 683 65
AK166198.1 - 6 - 7 862 65
BC006663.1 - 6 - 7 856 3498551 65
M24119.1 - 5 - 6 611 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000750.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gba

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
lipid metabolic process
sphingolipid metabolic process
glucosylceramide catabolic process
metabolic process
response to pH
termination of signal transduction
negative regulation of interleukin-6 production
regulation of water loss via skin
response to testosterone
positive regulation of protein dephosphorylation
negative regulation of MAP kinase activity
skin morphogenesis
response to estrogen
sphingosine biosynthetic process
ceramide biosynthetic process
response to glucocorticoid
cellular response to tumor necrosis factor
response to thyroid hormone
positive regulation of proteolysis involved in cellular protein catabolic process
glucosylceramidase activity
receptor binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0