This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00481339_g1
See other RGCC GE Assays ›
Gene Symbol:
Rgcc
Gene Name:
regulator of cell cycle
Gene Aliases:
1190002H23Rik, Rgc-32, Rgc32
Chromosome Location:
Chr.14: 79288750 - 79301635 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00481339_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgcc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025427.2 NP_079703.2 1 - 2 192 136
GenBank mRNA AF276981.1 - 1 - 2 204 136
AK004451.1 - 1 - 2 193 136
AK004705.1 - 1 - 2 188 136
BC049580.1 - 1 - 2 188 6741281 136
BC050935.1 - 1 - 2 181 6818687 136
BY083770.1 - 1 - 2 194 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002487.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF276981.1,AK004451.1,AK004705.1,BC049580.1,BC050935.1,NM_025427.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgcc

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
negative regulation of exit from mitosis
negative regulation of endothelial cell proliferation
positive regulation of extracellular matrix constituent secretion
complement activation
cell cycle
negative regulation of cell proliferation
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of angiogenesis
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
positive regulation of collagen biosynthetic process
negative regulation of blood vessel endothelial cell migration
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
negative regulation of cytokine secretion
positive regulation of cytokine secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of stress fiber assembly
regulation of cell cycle
positive regulation of cell cycle arrest
cellular response to hypoxia
mitotic cell cycle arrest
fibroblast activation
negative regulation of fibroblast growth factor production
positive regulation of extracellular matrix assembly
negative regulation of mitotic cell cycle phase transition
negative regulation of cell-cell adhesion mediated by cadherin
positive regulation of endothelial cell apoptotic process
protein binding
protein kinase binding
protein kinase activator activity
R-SMAD binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019104.mm.1 NM_025427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019104.mm.1 NM_025427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019104.mm.1 TC1400002487.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019103.mm.1 NM_025427.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019103.mm.1 NM_025427.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019103.mm.1 TC1400002487.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 802003 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0