Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000011940.mm.1 |
NM_025364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011940.mm.1 |
NM_025364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011940.mm.1 |
TC1000001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011946.mm.1 |
NM_025364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011946.mm.1 |
NM_025364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011946.mm.1 |
TC1000001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011944.mm.1 |
NM_025364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011944.mm.1 |
NM_025364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011944.mm.1 |
TC1000001624.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011942.mm.1 |
NM_025364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011942.mm.1 |
NM_025364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011942.mm.1 |
TC1000001624.mm.1 |
MTA TC |