This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00480584_m1
See other GPR180 GE Assays ›
Gene Symbol:
Gpr180
Gene Name:
G protein-coupled receptor 180
Gene Aliases:
AB041545, E130016I23Rik, Itr
Chromosome Location:
Chr.14: 118137101 - 118164232 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00480584_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr180
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021434.5 NP_067409.2 5 - 6 804 67
GenBank mRNA AB041545.1 - 5 - 6 796 67
AK053422.1 - 5 - 6 774 67
AK169908.1 - 5 - 6 803 67
AK170316.1 - 5 - 6 769 67
BC055726.1 - 5 - 6 780 67
BC069981.1 - 5 - 6 760 6336270 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001275.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr180

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
biological_process
response to pheromone
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009655.mm.1 NM_021434.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009655.mm.1 NM_021434.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400009655.mm.1 TC1400001275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400009656.mm.1 NM_021434.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009656.mm.1 NM_021434.5 Refseq
Affymetrix Mouse Clariom S Array PSR1400009656.mm.1 TC1400001275.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0