This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479880_m1
See other NAGK GE Assays ›
Gene Symbol:
Nagk
Gene Name:
N-acetylglucosamine kinase
Gene Aliases:
Gnk
Chromosome Location:
Chr.6: 83795031 - 83802556 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479880_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nagk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164187.1 NP_001157659.1 5 - 6 654 142
NM_019542.2 NP_062415.1 5 - 6 663 142
GenBank mRNA AJ242909.1 - 5 - 6 675 142
AJ242911.1 - 5 - 6 615 142
AK007240.1 - 5 - 6 658 142
AK155292.1 - 5 - 6 555 142
BC004689.1 - 5 - 6 541 3501470 142

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000941.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nagk

Gene Ontology Categories:

Function(s) Process(es)

N-acetylglucosamine metabolic process
phosphorylation
N-acetylneuraminate catabolic process
carbohydrate phosphorylation
nucleotide binding
ATP binding
N-acylmannosamine kinase activity
kinase activity
transferase activity
N-acetylglucosamine kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600007229.mm.1 NM_001164187.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007229.mm.1 NM_001164187.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007229.mm.1 NM_019542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007229.mm.1 NM_019542.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600007229.mm.1 TC0600000941.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007231.mm.1 NM_001164187.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007231.mm.1 NM_001164187.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007231.mm.1 NM_019542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007231.mm.1 NM_019542.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600007231.mm.1 TC0600000941.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0