Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100033513.mm.1 |
NM_018873.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033513.mm.1 |
NM_018873.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033513.mm.1 |
TC1100003646.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033512.mm.1 |
NM_018873.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033512.mm.1 |
NM_018873.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033512.mm.1 |
TC1100003646.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100033514.mm.1 |
NM_018873.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033514.mm.1 |
NM_018873.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100033514.mm.1 |
TC1100003646.mm.1 |
MTA TC |