This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00479407_m1
See other NELL2 GE Assays ›
Gene Symbol:
Nell2
Gene Name:
NEL-like 2
Gene Aliases:
A330108N19Rik, R75516, mel91
Chromosome Location:
Chr.15: 95219440 - 95528758 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00479407_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nell2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289653.1 NP_001276582.1 17 - 18 1850 78
GenBank mRNA AK035019.1 - 15 - 16 1703 78
AK039790.1 - 1914 78
AK048913.1 - 2121 78
BC051968.1 - 16 - 17 1818 6401465 78
U59230.1 - 17 - 18 1851 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

calcium-binding protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of growth
development of secondary female sexual characteristics
positive regulation of hormone secretion
neuron cellular homeostasis
protein homotrimerization
protein kinase C binding
calcium ion binding
heparin binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0