This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478587_m1
See other S100A3 GE Assays ›
Gene Symbol:
S100a3
Gene Name:
S100 calcium binding protein A3
Gene Aliases:
S100E
Chromosome Location:
Chr.3: 90560356 - 90602706 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478587_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011310.2 NP_035440.1 1 - 2 59 77
GenBank mRNA AF004941.1 - 1 - 2 59 77
AK133717.1 - 1 - 2 61 77
BC099893.1 - 1 - 2 56 40043723 77
BC100690.1 - 1 - 2 56 40043727 77
BC100696.1 - 1 - 2 56 40043722 77
BC100697.1 - 1 - 2 56 40043719 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000806.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF004941.1,AK133717.1,BC099893.1,BC100690.1,BC100696.1,BC100697.1,NM_011310.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100a3

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

calcium ion binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006284.mm.1 NM_011310.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006284.mm.1 NM_011310.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300006284.mm.1 TC0300000806.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300006285.mm.1 NM_011310.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006285.mm.1 NM_011310.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300006285.mm.1 TC0300000806.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 349958 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0