This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478121_m1
See other PER3 GE Assays ›
Gene Symbol:
Per3
Gene Name:
period circadian clock 3
Gene Aliases:
2810049O06Rik, mPer3
Chromosome Location:
Chr.4: 151003655 - 151044675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478121_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Per3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289877.1 NP_001276806.1 5 - 6 1074 66
NM_001289878.1 NP_001276807.1 6 - 7 1123 66
NM_011067.3 NP_035197.2 5 - 6 1074 66
GenBank mRNA AB013605.1 - 5 - 6 881 66
AF050182.1 - 5 - 6 998 66
AJ007377.1 - 5 - 6 881 66
AK036345.1 - 5 - 6 991 66
BC141032.1 - 5 - 6 672 9055644 66
BC145178.1 - 6 - 7 721 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001289878.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Per3

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
circadian rhythm
circadian regulation of gene expression
regulation of circadian sleep/wake cycle, sleep
rhythmic process
signal transducer activity
protein binding
kinase binding
ubiquitin protein ligase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0