This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00478093_g1
See other PADI4 GE Assays ›
Gene Symbol:
Padi4
Gene Name:
peptidyl arginine deiminase, type IV
Gene Aliases:
Pad4, Pdi4
Chromosome Location:
Chr.4: 140745508 - 140774239 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00478093_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Padi4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011061.2 NP_035191.2 12 - 13 1480 96
GenBank mRNA AB013850.1 - 12 - 13 1473 96
AK137625.1 - 12 - 13 1483 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003865.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Padi4

Gene Ontology Categories:

Function(s) Process(es)

immune system process
nucleosome assembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
protein citrullination
stem cell population maintenance
histone H3-R26 citrullination
histone citrullination
innate immune response
protein-arginine deiminase activity
calcium ion binding
hydrolase activity
arginine deiminase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400031882.mm.1 NM_011061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400031882.mm.1 NM_011061.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400031882.mm.1 TC0400003865.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400031884.mm.1 NM_011061.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400031884.mm.1 NM_011061.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400031884.mm.1 TC0400003865.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0