This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00477702_g1
See other HRC GE Assays ›
Gene Symbol:
Hrc
Gene Name:
histidine rich calcium binding protein
Gene Aliases:
-
Chromosome Location:
Chr.7: 45335269 - 45338973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00477702_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hrc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010473.2 NP_034603.2 3 - 4 2243 76
GenBank mRNA AF132218.1 - 3 - 4 2222 76
AF158597.1 - 4 - 5 2077 76
AI322629.1 - 3 - 4 194 76
BC141121.1 - 5 - 6 2062 9055733 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000840.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hrc

Gene Ontology Categories:

Function(s)

regulation of heart contraction
negative regulation of cytosolic calcium ion concentration
calcium ion homeostasis
negative regulation of calcium ion import into sarcoplasmic reticulum

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700007239.mm.1 NM_010473.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007239.mm.1 NM_010473.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700007239.mm.1 TC0700000840.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007238.mm.1 NM_010473.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007238.mm.1 NM_010473.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700007238.mm.1 TC0700000840.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0