This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00477067_g1
See other INPP5K GE Assays ›
Gene Symbol:
Inpp5k
Gene Name:
inositol polyphosphate 5-phosphatase K
Gene Aliases:
C62, Pps, SKIP
Chromosome Location:
Chr.11: 75630507 - 75648865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00477067_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Inpp5k
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008916.2 NP_032942.1 5 - 6 688 83
GenBank mRNA AF483522.1 - 5 - 6 608 83
AF483523.1 - 5 - 6 608 83
AK054436.1 - 5 - 6 707 83
AK078104.1 - 5 - 6 699 83
AK164290.1 - 5 - 6 677 83
BC014725.1 - 5 - 6 618 3969089 83
BC066112.1 - 5 - 6 688 30361046 83
U96724.1 - 5 - 6 719 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001076.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Inpp5k

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
negative regulation of protein phosphorylation
regulation of glycogen biosynthetic process
negative regulation of protein kinase activity
G-protein coupled receptor signaling pathway
negative regulation of peptidyl-threonine phosphorylation
negative regulation of glucose transport
dephosphorylation
response to insulin
cellular response to insulin stimulus
cellular response to hormone stimulus
negative regulation of peptidyl-serine phosphorylation
negative regulation of dephosphorylation
positive regulation of urine volume
glucose homeostasis
negative regulation of MAP kinase activity
negative regulation by host of viral transcription
negative regulation of glycogen biosynthetic process
negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of insulin receptor signaling pathway
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
negative regulation of stress fiber assembly
negative regulation of protein kinase B signaling
negative regulation of calcium ion transport
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
protein targeting to plasma membrane
negative regulation of protein targeting to membrane
ruffle assembly
negative regulation of glycogen (starch) synthase activity
positive regulation of renal water transport
inositol-polyphosphate 5-phosphatase activity
vasopressin receptor activity
inositol bisphosphate phosphatase activity
hydrolase activity
phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
phosphatidylinositol trisphosphate phosphatase activity
phosphatidylinositol phosphate 5-phosphatase activity
inositol trisphosphate phosphatase activity
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100009812.mm.1 NM_008916.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100009812.mm.1 NM_008916.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100009812.mm.1 TC1100001076.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0