This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00476840_g1
See other PIP5K1A GE Assays ›
Gene Symbol:
Pip5k1a
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha
Gene Aliases:
PI4P5K-I[a], PIP5K1-alpha, PIP5KIalpha, Pipk5a
Chromosome Location:
Chr.3: 95058530 - 95106930 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00476840_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pip5k1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293707.1 NP_001280636.1 12 - 13 1944 61
NM_008847.3 NP_032873.2 12 - 13 1941 61
GenBank mRNA AK098097.1 - 12 - 13 1898 61
AK150898.1 - 12 - 13 1901 61
AK151984.1 - 12 - 13 1901 61
AK158062.1 - 12 - 13 1886 61
BC003763.1 - 9 - 10 998 3591003 61
BC031774.1 - 12 - 13 1869 4503697 61
D86177.1 - 12 - 13 1731 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002463.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pip5k1a

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid biosynthetic process
fibroblast migration
phosphorylation
actin cytoskeleton reorganization
phosphatidylinositol metabolic process
phosphatidylinositol phosphorylation
focal adhesion assembly
cell chemotaxis
protein targeting to plasma membrane
activation of GTPase activity
ruffle assembly
nucleotide binding
protein binding
ATP binding
kinase activity
phosphatidylinositol phosphate kinase activity
1-phosphatidylinositol-4-phosphate 5-kinase activity
transferase activity
kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300019002.mm.1 NM_008847.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300019002.mm.1 TC0300002463.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0