This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00476810_g1
See other PIP5K1C GE Assays ›
Gene Symbol:
Pip5k1c
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase, type 1 gamma
Gene Aliases:
AI115456, AI835305, Pip5kIgamma
Chromosome Location:
Chr.10: 81292962 - 81319974 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00476810_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pip5k1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146687.2 NP_001140159.1 9 - 10 1304 71
NM_001293646.1 NP_001280575.1 9 - 10 1304 71
NM_001293647.1 NP_001280576.1 9 - 10 1317 71
NM_008844.3 NP_032870.2 9 - 10 1304 71
GenBank mRNA AB006916.1 - 9 - 10 1287 71
AK014403.1 - 9 - 10 1270 71
AK041402.1 - 9 - 10 1290 71
AK122319.2 - 9 - 10 1262 71
AK154816.1 - 9 - 10 1296 71
AK155371.1 - 9 - 10 1280 71
AK170286.1 - 9 - 10 1296 71
AK171576.1 - 9 - 10 1278 71
AK203613.1 - 1 - 2 51 71
BC019138.1 - 9 - 10 1253 5041431 71
BC020365.1 - 9 - 10 1266 71
BC094665.1 - 9 - 10 1276 30356181 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000892.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pip5k1c

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
endocytosis
phagocytosis
chemotaxis
cytoskeletal anchoring at plasma membrane
cell adhesion
axonogenesis
phosphorylation
phosphatidylinositol metabolic process
phosphatidylinositol phosphorylation
platelet aggregation
nucleotide binding
protein binding
ATP binding
kinase activity
phosphatidylinositol phosphate kinase activity
1-phosphatidylinositol-4-phosphate 5-kinase activity
transferase activity
talin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000007010.mm.1 NM_001146687.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000007010.mm.1 NM_008844.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000007010.mm.1 TC1000000892.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000007009.mm.1 NM_001146687.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000007009.mm.1 NM_008844.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000007009.mm.1 TC1000000892.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0