This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00476077_m1
See other BACH1 GE Assays ›
Gene Symbol:
Bach1
Gene Name:
BTB and CNC homology 1
Gene Aliases:
6230421P05Rik, AI323795
Chromosome Location:
Chr.16: 87698954 - 87733346 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00476077_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bach1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007520.2 NP_031546.1 1 - 2 103 109
GenBank mRNA AK140765.1 - 1 - 2 104 109
AK140903.1 - 1 - 2 104 109
D86603.1 - 1 - 2 113 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000958.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK140765.1,AK140903.1,D86603.1,NM_007520.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bach1

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
regulation of transcription involved in G2/M transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to hypoxia
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600007518.mm.1 NM_007520.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007518.mm.1 NM_007520.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007518.mm.1 TC1600000958.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007515.mm.1 NM_007520.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007515.mm.1 NM_007520.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007515.mm.1 TC1600000958.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600007516.mm.1 NM_007520.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007516.mm.1 NM_007520.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600007516.mm.1 TC1600000958.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 552621 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0