This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00475831_m1
See other ALPL GE Assays ›
Gene Symbol:
Alpl
Gene Name:
alkaline phosphatase, liver/bone/kidney
Gene Aliases:
APTNAP, Akp-2, Akp2, TNAP, TNSALP
Chromosome Location:
Chr.4: 137741731 - 137796384 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00475831_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Alpl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007431.3 NP_031457.2 1 - 2 144 141
GenBank mRNA AK076459.1 - 1 - 2 80 140
AK161780.1 - 1 - 2 80 141
AK164002.1 - 1 - 2 138 140
AK166732.1 - 1 - 2 78 140
AK167023.1 - 1 - 2 80 140
AK167481.1 - 1 - 2 79 141
BC065175.1 - 1 - 2 67 6807509 141
J02980.1 - 1 - 2 83 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003814.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK161780.1,AK164002.1,AK167023.1,AK167481.1,BC065175.1,J02980.1,NM_007431.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Alpl

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
endochondral ossification
developmental process involved in reproduction
metabolic process
dephosphorylation
response to antibiotic
response to glucocorticoid
cellular response to organic cyclic compound
catalytic activity
alkaline phosphatase activity
protein binding
pyrophosphatase activity
hydrolase activity
phosphatase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400031451.mm.1 NM_001287172.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400031451.mm.1 NM_007431.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400031451.mm.1 TC0400003814.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400031460.mm.1 NM_007431.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400031460.mm.1 TC0400003814.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400031461.mm.1 NM_007431.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400031461.mm.1 TC0400003814.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400031452.mm.1 NM_001287172.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400031452.mm.1 NM_007431.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400031452.mm.1 TC0400003814.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 525289 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0