This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00475710_m1
See other ACVR1B GE Assays ›
Gene Symbol:
Acvr1b
Gene Name:
activin A receptor, type 1B
Gene Aliases:
6820432J04, ActR-IB, ActRIB, Acvrlk4, Alk4, SKR2
Chromosome Location:
Chr.15: 101174125 - 101212601 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00475710_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acvr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007395.3 NP_031421.1 5 - 6 998 83
GenBank mRNA AK154578.1 - 1021 83
AK157910.1 - 5 - 6 999 83
BC059832.1 - 991 83
BC145775.1 - 5 - 6 1051 40131078 83
BC145777.1 - 5 - 6 1051 40131080 83
Z31663.1 - 5 - 6 987 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001062.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acvr1b

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
in utero embryonic development
hair follicle development
regulation of transcription, DNA-templated
protein phosphorylation
signal transduction
cell surface receptor signaling pathway
transmembrane receptor protein serine/threonine kinase signaling pathway
mesoderm development
embryo development
regulation of signal transduction
positive regulation of gene expression
negative regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
negative regulation of cell growth
activin receptor signaling pathway
positive regulation of activin receptor signaling pathway
intracellular signal transduction
nodal signaling pathway
positive regulation of erythrocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
extrinsic apoptotic signaling pathway
positive regulation of trophoblast cell migration
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
activin receptor activity, type I
transferase activity
transferase activity, transferring phosphorus-containing groups
activin-activated receptor activity
growth factor binding
inhibin binding
SMAD binding
metal ion binding
activin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008461.mm.1 NM_007395.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008461.mm.1 NM_007395.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500008461.mm.1 TC1500001062.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008462.mm.1 NM_007395.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008462.mm.1 NM_007395.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500008462.mm.1 TC1500001062.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0