Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200014054.mm.1 |
NM_001267625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014054.mm.1 |
NM_001267626.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014054.mm.1 |
NM_058212.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014054.mm.1 |
TC1200002045.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014057.mm.1 |
NM_001267625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014057.mm.1 |
NM_001267626.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014057.mm.1 |
NM_058212.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014057.mm.1 |
TC1200002045.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200014061.mm.1 |
NM_001267625.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014061.mm.1 |
NM_001267626.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014061.mm.1 |
NM_058212.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200014061.mm.1 |
TC1200002045.mm.1 |
MTA TC |