This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00475389_m1
See other NECAB2 GE Assays ›
Gene Symbol:
Necab2
Gene Name:
N-terminal EF-hand calcium binding protein 2
Gene Aliases:
Efcbp2
Chromosome Location:
Chr.8: 119446719 - 119472640 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00475389_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Necab2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_054095.2 NP_473436.1 8 - 9 1138 60
GenBank mRNA AF411253.1 - 8 - 9 1132 60
AK035168.1 - 6 - 7 511 60
AK082727.1 - 8 - 9 748 60
BC107384.1 - 9 - 10 733 60
BC107385.1 - 9 - 10 697 60
BC115857.1 - 9 - 10 670 40057629 60
BC115963.1 - 9 - 10 658 40057628 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001421.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Necab2

Gene Ontology Categories:

Process(es)

calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800011559.mm.1 NM_054095.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011559.mm.1 NM_054095.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011559.mm.1 TC0800001421.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800011558.mm.1 NM_054095.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800011558.mm.1 NM_054095.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800011558.mm.1 TC0800001421.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0