Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800011553.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011553.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011553.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011544.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011544.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011544.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011540.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011540.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011540.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011542.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011542.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011542.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011550.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011550.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011550.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011545.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011545.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011545.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011546.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011546.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011546.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011548.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011548.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011548.mm.1 |
TC0800001421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011549.mm.1 |
NM_054095.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011549.mm.1 |
NM_054095.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011549.mm.1 |
TC0800001421.mm.1 |
MTA TC |