This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00474097_g1
See other CD163 GE Assays ›
Gene Symbol:
Cd163
Gene Name:
CD163 antigen
Gene Aliases:
CD163v2, CD163v3
Chromosome Location:
Chr.6: 124304631 - 124330529 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00474097_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd163
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170395.1 NP_001163866.1 12 - 13 3183 72
NM_053094.2 NP_444324.2 12 - 13 3183 72
GenBank mRNA AF274883.1 - 12 - 13 3178 72
AK032012.1 - 2 - 3 505 72
BC144848.1 - 12 - 13 3134 9053396 72
BC145793.1 - 12 - 13 3134 40131096 72
DQ058616.1 - 12 - 13 3139 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001418.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd163

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
acute-phase response
inflammatory response
scavenger receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600011396.mm.1 NM_001170395.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600011396.mm.1 NM_001170395.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600011396.mm.1 NM_053094.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600011396.mm.1 NM_053094.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600011396.mm.1 TC0600001418.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0