This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00474036_mH
See other LRP8 GE Assays ›
Gene Symbol:
Lrp8
Gene Name:
low density lipoprotein receptor-related protein 8, apolipoprotein e receptor
Gene Aliases:
4932703M08Rik, AA921429, AI848122, ApoER2, Lr8b
Chromosome Location:
Chr.4: 107802259 - 107876840 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00474036_mH
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lrp8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080926.1 NP_001074395.1 16 - 17 2381 89
NR_033496.1 17 - 18 2759 89
GenBank mRNA AJ312058.1 - 16 - 17 2227 89
AK030143.1 - 13 - 14 1891 89
AK038580.1 - 15 - 16 2418 89
BC127061.1 - 17 - 18 2134 89
D85463.1 - 17 - 18 2757 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001135.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

apolipoprotein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
endocytosis
receptor-mediated endocytosis
ammon gyrus development
hippocampus development
layer formation in cerebral cortex
positive regulation of CREB transcription factor activity
reelin-mediated signaling pathway
positive regulation of protein kinase activity
positive regulation of peptidyl-tyrosine phosphorylation
modulation of synaptic transmission
positive regulation of dendritic spine morphogenesis
positive regulation of protein tyrosine kinase activity
positive regulation of dendrite development
glycoprotein binding
low-density lipoprotein receptor activity
calcium ion binding
protein binding
high-density lipoprotein particle binding
kinesin binding
very-low-density lipoprotein particle receptor activity
apolipoprotein binding
reelin receptor activity
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400007953.mm.1 NM_001080926.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007953.mm.1 NM_001080926.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007953.mm.1 NR_033496.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007953.mm.1 NR_033496.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400007953.mm.1 TC0400001135.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0