This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00473833_m1
See other MPV17L GE Assays ›
Gene Symbol:
Mpv17l
Gene Name:
Mpv17 transgene, kidney disease mutant-like
Gene Aliases:
M-LP
Chromosome Location:
Chr.16: 13903161 - 13949619 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00473833_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpv17l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289561.1 NP_001276490.1 1 - 2 359 57
NM_033564.3 NP_291042.2 1 - 2 359 57
GenBank mRNA AF305634.1 - 1 - 2 329 57
BC094450.1 - 1 - 2 326 4973897 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000168.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mpv17l

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of hydrogen peroxide biosynthetic process
reactive oxygen species metabolic process
negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0