This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471750_g1
See other MFSD10 GE Assays ›
Gene Symbol:
Mfsd10
Gene Name:
major facilitator superfamily domain containing 10
Gene Aliases:
0610009O03Rik, Tetran
Chromosome Location:
Chr.5: 34633644 - 34637215 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471750_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mfsd10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026660.2 NP_080936.1 10 - 11 1323 69
GenBank mRNA AK003763.1 - 5 - 6 591 69
AK018736.1 - 10 - 11 1336 69
AK089055.1 - 10 - 11 1340 69
AK144407.1 - 1 - 2 43 69
BC023012.1 - 10 - 11 1323 5376230 69
BC064775.1 - 10 - 11 1221 30023877 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002239.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mfsd10

Gene Ontology Categories:

Function(s) Process(es)

transport
apoptotic process
transmembrane transport
transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500020474.mm.1 NM_026660.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500020474.mm.1 NM_026660.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500020474.mm.1 TC0500002239.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0